Product Details

SNP ID
rs1045018
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.5:132369935 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CACGCGCAAAGCCCGCCGCGTTCCC[A/C]GACCCCAGGCCGCGCTCTGTGGGCC
Phenotype
MIM: 603377
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC553103 PubMed Links

Gene Details

Gene
LOC553103
Gene Name
uncharacterized LOC553103
There are no transcripts associated with this gene.

Gene
MIR3936
Gene Name
microRNA 3936
There are no transcripts associated with this gene.

Gene
SLC22A5
Gene Name
solute carrier family 22 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308122.1 227 UTR 5 NP_001295051.1
NM_003060.3 227 UTR 5 NP_003051.1
XM_011543590.2 227 Intron XP_011541892.1
XM_017009778.1 227 Intron XP_016865267.1

View Full Product Details