Product Details

SNP ID
rs7597243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:159935398 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTTGTTTCTCCCATTTCTTCTA[A/C]ATTTATCAATTGGAATTTACTGGGG
Phenotype
MIM: 604939
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLA2R1 PubMed Links

Gene Details

Gene
PLA2R1
Gene Name
phospholipase A2 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007267.2 Intron NP_001007268.1
NM_001195641.1 Intron NP_001182570.1
NM_007366.4 Intron NP_031392.3
XM_005246392.3 Intron XP_005246449.1
XM_011510820.2 Intron XP_011509122.1
XM_017003598.1 Intron XP_016859087.1
XM_017003599.1 Intron XP_016859088.1
XM_017003600.1 Intron XP_016859089.1
XM_017003601.1 Intron XP_016859090.1
XM_017003602.1 Intron XP_016859091.1

View Full Product Details