Product Details

SNP ID
rs11554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124619376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTAGAGCGGCTGGAGTGCAAGC[C/T]ACAGCCTGTGCGGGGAGGCCAGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM53B PubMed Links

Gene Details

Gene
FAM53B
Gene Name
family with sequence similarity 53 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014661.3 5658 UTR 3 NP_055476.3
Gene
LHPP
Gene Name
phospholysine phosphohistidine inorganic pyrophosphate phosphatase
There are no transcripts associated with this gene.

View Full Product Details