Product Details
- SNP ID
-
rs4924445
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40281212 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCTGGCTGTAGCCTACGGAACCA[A/G]TCAATCGACACCTCTAGACCTAGGC
- Phenotype
-
MIM: 608110
MIM: 604114
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD63
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112189073,rs77350758] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD63
- Gene Name
- ankyrin repeat domain 63
- Gene
- BUB1B-PAK6
- Gene Name
- BUB1B-PAK6 readthrough
There are no transcripts associated with this gene.
- Gene
- PAK6
- Gene Name
- p21 (RAC1) activated kinase 6
There are no transcripts associated with this gene.
- Gene
- PLCB2
- Gene Name
- phospholipase C beta 2
There are no transcripts associated with this gene.
View Full Product Details