Product Details

SNP ID
rs7022141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:76773150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCATGCTTATTTCCCAAGTTAA[A/T]TTTACTTCACCAAAGCGGTCTTTTG
Phenotype
MIM: 604845 MIM: 610691
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PCA3 PubMed Links

Gene Details

Gene
PCA3
Gene Name
prostate cancer associated 3 (non-protein coding)
There are no transcripts associated with this gene.

Gene
PRUNE2
Gene Name
prune homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308047.1 Intron NP_001294976.1
NM_001308048.1 Intron NP_001294977.1
NM_001308049.1 Intron NP_001294978.1
NM_001308050.1 Intron NP_001294979.1
NM_001308051.1 Intron NP_001294980.1
NM_015225.2 Intron NP_056040.2
XM_005251745.1 Intron XP_005251802.1
XM_005251746.1 Intron XP_005251803.1
XM_005251748.1 Intron XP_005251805.1
XM_005251750.1 Intron XP_005251807.1
XM_005251751.1 Intron XP_005251808.1
XM_005251754.1 Intron XP_005251811.1
XM_006716982.1 Intron XP_006717045.1
XM_006716983.1 Intron XP_006717046.1
XM_006716984.2 Intron XP_006717047.1
XM_006716985.1 Intron XP_006717048.1
XM_006716986.1 Intron XP_006717049.1
XM_011518323.1 Intron XP_011516625.1
XM_011518326.2 Intron XP_011516628.1
XM_011518327.1 Intron XP_011516629.1
XM_011518328.2 Intron XP_011516630.1
XM_017014345.1 Intron XP_016869834.1
XM_017014346.1 Intron XP_016869835.1
XM_017014347.1 Intron XP_016869836.1
XM_017014348.1 Intron XP_016869837.1
XM_017014349.1 Intron XP_016869838.1
XM_017014350.1 Intron XP_016869839.1
XM_017014351.1 Intron XP_016869840.1
XM_017014352.1 Intron XP_016869841.1
XM_017014353.1 Intron XP_016869842.1
XM_017014354.1 Intron XP_016869843.1
XM_017014355.1 Intron XP_016869844.1
XM_017014356.1 Intron XP_016869845.1
XM_017014357.1 Intron XP_016869846.1
XM_017014358.1 Intron XP_016869847.1
XM_017014359.1 Intron XP_016869848.1

View Full Product Details