Product Details

SNP ID
rs11635570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65002565 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTTTTGTAATTCAAGTAAAAAA[A/G]AAGCTGTTTTTTAACTACTGTTACA
Phenotype
MIM: 611766
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTFMT PubMed Links

Gene Details

Gene
MTFMT
Gene Name
mitochondrial methionyl-tRNA formyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139242.3 1835 UTR 3 NP_640335.2
XM_005254158.4 1835 UTR 3 XP_005254215.2

View Full Product Details