Product Details

SNP ID
rs10467951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75731380 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCTCATCTCATGCACAGATACG[C/T]GCCCACCCAGGCTCACTGGCGCCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNM1P35 PubMed Links

Gene Details

Gene
DNM1P35
Gene Name
dynamin 1 pseudogene 35
There are no transcripts associated with this gene.

Gene
ODF3L1
Gene Name
outer dense fiber of sperm tails 3 like 1
There are no transcripts associated with this gene.

View Full Product Details