Product Details

SNP ID
rs9921035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45254462 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATACTTACCATTGTGCTATAGTT[A/G]CCTACAGTATTAAGTACAGAAACAT
Phenotype
MIM: 606208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928414 PubMed Links

Gene Details

Gene
LOC101928414
Gene Name
uncharacterized LOC101928414
There are no transcripts associated with this gene.

Gene
SLC28A2
Gene Name
solute carrier family 28 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004212.3 Intron NP_004203.2
XM_011522198.2 Intron XP_011520500.1
XM_011522199.1 Intron XP_011520501.1
XM_011522200.2 Intron XP_011520502.1
XM_011522201.2 Intron XP_011520503.1
XM_011522202.2 Intron XP_011520504.1

View Full Product Details