Product Details
- SNP ID
-
rs6767632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:132737072 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGTTTTCCCAACACTATCAACTA[A/G]ACTGTCCTTTCCCTATTGTGTGCAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPHP3-AS1
PubMed Links
Gene Details
- Gene
- NPHP3-AS1
- Gene Name
- NPHP3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details