Product Details

SNP ID
rs34988350
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:28506084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGCAGTGGTCGCTTCTTCTCCTT[A/G]GTAAGTGTGATCCTTGGTAAGTGTG
Phenotype
MIM: 608726 MIM: 179710 MIM: 603238 MIM: 180645 MIM: 603239
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHACTR4 PubMed Links

Gene Details

Gene
PHACTR4
Gene Name
phosphatase and actin regulator 4
There are no transcripts associated with this gene.

Gene
RCC1
Gene Name
regulator of chromosome condensation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048194.2 142 Intron NP_001041659.1
NM_001048195.2 142 Intron NP_001041660.1
NM_001048199.2 142 UTR 5 NP_001041664.1
NM_001269.4 142 Intron NP_001260.1
Gene
SNHG3
Gene Name
small nucleolar RNA host gene 3
There are no transcripts associated with this gene.

Gene
SNORA73A
Gene Name
small nucleolar RNA, H/ACA box 73A
There are no transcripts associated with this gene.

Gene
SNORA73B
Gene Name
small nucleolar RNA, H/ACA box 73B
There are no transcripts associated with this gene.

View Full Product Details