Product Details

SNP ID
rs6499882
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57535535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCTGAGGGGCACAATGCCCCCG[A/G]GAGCACCTACACGATGGGGTCTGCA
Phenotype
MIM: 616965
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRG5 PubMed Links

Gene Details

Gene
ADGRG5
Gene Name
adhesion G protein-coupled receptor G5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304376.2 Intron NP_001291305.1
NM_001318481.1 Intron NP_001305410.1
NM_153837.3 Intron NP_722579.1
XM_011522949.2 Intron XP_011521251.1
XM_011522950.2 Intron XP_011521252.1
Gene
CCDC102A
Gene Name
coiled-coil domain containing 102A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033212.3 Intron NP_149989.2
XM_011523469.2 Intron XP_011521771.1

View Full Product Details