Product Details

SNP ID
rs152653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:49379165 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTTCATCTATAAAATGGGACAA[C/T]AACAATACCTGCCTCATAGTATGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf78 PubMed Links

Gene Details

Gene
C16orf78
Gene Name
chromosome 16 open reading frame 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144602.3 Intron NP_653203.1

View Full Product Details