Product Details

SNP ID
rs3108290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185217384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTTTGATACAATTCTCATTA[A/T]TTATCATTACAATAGGTTCAAGAAT
Phenotype
MIM: 616047
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFAP97 PubMed Links

Gene Details

Gene
CFAP97
Gene Name
cilia and flagella associated protein 97
There are no transcripts associated with this gene.

Gene
SNX25
Gene Name
sorting nexin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317781.1 Intron NP_001304710.1
XM_005263267.4 Intron XP_005263324.1
XM_011532302.2 Intron XP_011530604.1
XM_011532303.2 Intron XP_011530605.1
XM_011532304.2 Intron XP_011530606.1
XM_011532305.2 Intron XP_011530607.1
XM_011532306.2 Intron XP_011530608.1
XM_011532307.2 Intron XP_011530609.1
XM_011532308.2 Intron XP_011530610.1
XM_017008680.1 Intron XP_016864169.1
XM_017008681.1 Intron XP_016864170.1
XM_017008682.1 Intron XP_016864171.1
XM_017008683.1 Intron XP_016864172.1
XM_017008684.1 Intron XP_016864173.1
XM_017008685.1 Intron XP_016864174.1
XM_017008686.1 Intron XP_016864175.1
XM_017008687.1 Intron XP_016864176.1

View Full Product Details