Product Details
- SNP ID
-
rs12998798
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:49920617 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTGGCACAACCCTCTTCCTTCCT[C/G]CTTTTCAATTTGTCAATAAATATTT
- Phenotype
-
MIM: 600565
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NRXN1
PubMed Links
Gene Details
- Gene
- NRXN1
- Gene Name
- neurexin 1
View Full Product Details