Product Details

SNP ID
rs861816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21701292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTCATCACTCTTTTGGCAAAAA[A/C]CCCCCCCAAGTCTATACTATGAACT
Phenotype
MIM: 607588 MIM: 608082
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPIL2 PubMed Links

Gene Details

Gene
PPIL2
Gene Name
peptidylprolyl isomerase like 2
There are no transcripts associated with this gene.

Gene
YPEL1
Gene Name
yippee like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013313.4 Intron NP_037445.1

View Full Product Details