Product Details
- SNP ID
-
rs11675922
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:9424694 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCACACCTGTCATGTCTTCCTG[C/T]GATGAGCAAACACTGAGCAAGTGTC
- Phenotype
-
MIM: 606029
MIM: 607153
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CPSF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116446789] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CPSF3
- Gene Name
- cleavage and polyadenylation specific factor 3
- Gene
- ITGB1BP1
- Gene Name
- integrin subunit beta 1 binding protein 1
There are no transcripts associated with this gene.
View Full Product Details