Product Details

SNP ID
rs10214312
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:132486174 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATTCAGTGCCAGGTGGAGTTCTGA[G/T]CATCTTTTCTCTCTCAGGAAGCCCT
Phenotype
MIM: 147575
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IRF1 PubMed Links

Gene Details

Gene
IRF1
Gene Name
interferon regulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002198.2 Intron NP_002189.1
XM_011543378.2 Intron XP_011541680.1
XM_011543379.2 Intron XP_011541681.1

View Full Product Details