Product Details

SNP ID
rs2577539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128965518 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGCCCTGAGGCCACGGACCGAG[A/G]CGTGGTGCTGAGCCCCTGCGCGGTT
Phenotype
MIM: 604196
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC27A6 PubMed Links

Gene Details

Gene
SLC27A6
Gene Name
solute carrier family 27 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017372.2 392 UTR 5 NP_001017372.1
NM_001317984.1 392 UTR 5 NP_001304913.1
NM_014031.4 392 UTR 5 NP_054750.1

View Full Product Details