Product Details

SNP ID
rs6828570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55431595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACATTAATATCCAGTGTTTACCC[C/G]CTTCCAACAGGGACTCCTGTGGGAC
Phenotype
MIM: 601851 MIM: 614726
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLOCK PubMed Links

Gene Details

Gene
CLOCK
Gene Name
clock circadian regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267843.1 6373 UTR 3 NP_001254772.1
NM_004898.3 6373 UTR 3 NP_004889.1
XM_005265787.2 6373 UTR 3 XP_005265844.1
XM_011534409.2 6373 UTR 3 XP_011532711.1
XM_011534410.2 6373 UTR 3 XP_011532712.1
XM_011534411.2 6373 UTR 3 XP_011532713.1
XM_017008854.1 6373 UTR 3 XP_016864343.1
XM_017008855.1 6373 UTR 3 XP_016864344.1
Gene
TMEM165
Gene Name
transmembrane protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018475.4 6373 Intron NP_060945.2
XM_011534394.2 6373 Intron XP_011532696.1
XM_017008412.1 6373 Intron XP_016863901.1

View Full Product Details