Product Details

SNP ID
rs2298265
Assay Type
Validated
NCBI dbSNP Submissions
72
Location
Chr.1:151286567 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TCATTGTCAAGAACACTGTGTGTCC[C/T]GAGCAGTCTGAGGCCCTGGCTGGAG
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507670 PubMed Links

Gene Details

Gene
LOC100507670
Gene Name
uncharacterized LOC100507670
There are no transcripts associated with this gene.

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
There are no transcripts associated with this gene.

Gene
ZNF687
Gene Name
zinc finger protein 687
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304763.1 578 Silent Mutation CCC,CCT P92P NP_001291692.1
NM_001304764.1 578 Silent Mutation CCC,CCT P92P NP_001291693.1
NM_020832.2 578 Silent Mutation CCC,CCT P92P NP_065883.1
XM_005245366.3 578 Silent Mutation CCC,CCT P101P XP_005245423.1
XM_011509811.2 578 Silent Mutation CCC,CCT P92P XP_011508113.1
XM_011509812.2 578 Silent Mutation CCC,CCT P92P XP_011508114.1
XM_011509813.2 578 Silent Mutation CCC,CCT P92P XP_011508115.1

View Full Product Details