Product Details

SNP ID
rs4246525
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:151285263 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAGAATATGAATTGATTCCATG[C/T]GGACATGAATGAGTCCTTAAACCTA
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507670 PubMed Links

Gene Details

Gene
LOC100507670
Gene Name
uncharacterized LOC100507670
There are no transcripts associated with this gene.

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
There are no transcripts associated with this gene.

Gene
ZNF687
Gene Name
zinc finger protein 687
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304763.1 142 UTR 5 NP_001291692.1
NM_001304764.1 142 Intron NP_001291693.1
NM_020832.2 142 Intron NP_065883.1
XM_005245366.3 142 Intron XP_005245423.1
XM_011509811.2 142 Intron XP_011508113.1
XM_011509812.2 142 Intron XP_011508114.1
XM_011509813.2 142 Intron XP_011508115.1

View Full Product Details