Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001201478.1 | Intron | NP_001188407.1 | |||
NM_006893.2 | Intron | NP_008824.2 | |||
XM_005277509.3 | Intron | XP_005277566.1 | |||
XM_006711196.3 | Intron | XP_006711259.1 | |||
XM_011509257.2 | Intron | XP_011507559.1 | |||
XM_011509258.2 | Intron | XP_011507560.1 | |||
XM_017000530.1 | Intron | XP_016856019.1 | |||
XM_017000531.1 | Intron | XP_016856020.1 | |||
XM_017000532.1 | Intron | XP_016856021.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182663.3 | Intron | NP_872604.1 | |||
NM_182664.3 | Intron | NP_872605.1 | |||
NM_182665.3 | Intron | NP_872606.1 |