Product Details
- SNP ID
-
rs2257790
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:66135463 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AATATTATATTAATCAAGTACTAAC[C/T]CTGGTTTGAATGGGTAAATGCAGAA
- Phenotype
-
MIM: 606121
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CRCP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11560207] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRCP
- Gene Name
- CGRP receptor component
View Full Product Details