Product Details

SNP ID
rs2050694
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:52631533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTATTCTGAATAGGGGTTCATAG[C/G]CTTCATCAGACTGCCAAGGGGGACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM159A PubMed Links

Gene Details

Gene
FAM159A
Gene Name
family with sequence similarity 159 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042693.2 96 Intron NP_001036158.1
XM_011541377.2 96 UTR 5 XP_011539679.1
XM_011541378.2 96 UTR 5 XP_011539680.1

View Full Product Details