Product Details
- SNP ID
-
rs11978020
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:64240633 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGGCGAGGTCCTAAATCTCACAC[A/G]TAAATGCAATCCAGACTTGGAATTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF679
PubMed Links
Gene Details
- Gene
- ZNF679
- Gene Name
- zinc finger protein 679
View Full Product Details