Product Details

SNP ID
rs12403328
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:32121404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATTTAAATTTGTGCATTTTGAG[G/T]ATTCTCTGACCTCCAGGGAAGTTGT
Phenotype
MIM: 610563
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KPNA6 PubMed Links

Gene Details

Gene
KPNA6
Gene Name
karyopherin subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012316.4 Intron NP_036448.1
XM_005270711.3 Intron XP_005270768.1

View Full Product Details