Product Details

SNP ID
rs11211383
Assay Type
Validated
NCBI dbSNP Submissions
70
Location
Chr.1:38016684 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GATCATTTTCTCATAGTGGCTTTGA[G/T]TTGGCTCTGCCTTGGTGTTTGAGCC
Phenotype
MIM: 609440
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UTP11 PubMed Links

Gene Details

Gene
UTP11
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016037.3 Intron NP_057121.2
XM_011541555.2 Intron XP_011539857.1

View Full Product Details