Product Details

SNP ID
rs10773517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:128274154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCAAGACAATGGATTCCAGTTA[C/T]TCTCCCAGAGAGACTGATTTCTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM132C PubMed Links

Gene Details

Gene
TMEM132C
Gene Name
transmembrane protein 132C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136103.2 Intron NP_001129575.2
XM_011538998.2 Intron XP_011537300.1

View Full Product Details