Product Details

SNP ID
rs12126083
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:42821439 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTATCTGTTTTTCTCACTCATA[A/G]AGTTAGAAGTCTAAAAAGAACATTT
Phenotype
MIM: 609017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017922.1 Intron NP_001017922.1
NM_018538.3 Intron NP_061008.2
XM_006710313.3 Intron XP_006710376.1
XM_011540570.2 Intron XP_011538872.1
XM_011540571.2 Intron XP_011538873.1
Gene
SVBP
Gene Name
small vasohibin binding protein
There are no transcripts associated with this gene.

View Full Product Details