Product Details

SNP ID
rs6998654
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:22370009 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGTCCCACAGATCAATTGGGCAAT[C/T]TTAGGGAAAACAAACTTTTTTCTAC
Phenotype
MIM: 608736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC39A14 PubMed Links

Gene Details

Gene
SLC39A14
Gene Name
solute carrier family 39 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128431.2 Intron NP_001121903.1
NM_001135153.1 Intron NP_001128625.1
NM_001135154.1 Intron NP_001128626.1
NM_015359.4 Intron NP_056174.2
XM_005273465.2 Intron XP_005273522.1
XM_005273466.4 Intron XP_005273523.1
XM_006716323.2 Intron XP_006716386.1
XM_006716324.2 Intron XP_006716387.1
XM_011544478.2 Intron XP_011542780.1
XM_017013292.1 Intron XP_016868781.1
XM_017013293.1 Intron XP_016868782.1
XM_017013294.1 Intron XP_016868783.1
XM_017013295.1 Intron XP_016868784.1

View Full Product Details