Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145159.2 | 2172 | UTR 3 | NP_001138631.1 | ||
NM_001172562.1 | 2172 | UTR 3 | NP_001166033.1 | ||
NM_018250.3 | 2172 | UTR 3 | NP_060720.2 | ||
XM_006716356.2 | 2172 | UTR 3 | XP_006716419.1 | ||
XM_006716357.3 | 2172 | Intron | XP_006716420.1 | ||
XM_011544573.1 | 2172 | UTR 3 | XP_011542875.1 | ||
XM_011544574.1 | 2172 | UTR 3 | XP_011542876.1 | ||
XM_011544575.2 | 2172 | UTR 3 | XP_011542877.1 | ||
XM_011544576.1 | 2172 | UTR 3 | XP_011542878.1 |