Product Details

SNP ID
rs12736571
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:78895958 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACATAGTTTCATGAAAGAACACAG[A/G]AGACTGCACCTTAAATAGGGATTTC
Phenotype
MIM: 616419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRL4 PubMed Links

Gene Details

Gene
ADGRL4
Gene Name
adhesion G protein-coupled receptor L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022159.3 Intron NP_071442.2

View Full Product Details