Product Details

SNP ID
rs2800584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:15168498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATTTTCCAGTTAGCCACAAAGA[C/T]CTTTTTAAAAATTATTTTTAATGTC
Phenotype
MIM: 613574
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC39B PubMed Links

Gene Details

Gene
TTC39B
Gene Name
tetratricopeptide repeat domain 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168339.1 Intron NP_001161811.1
NM_001168340.1 Intron NP_001161812.1
NM_001168341.1 Intron NP_001161813.1
NM_001168342.1 Intron NP_001161814.1
NM_152574.2 Intron NP_689787.2
XM_011517732.2 Intron XP_011516034.1
XM_017014310.1 Intron XP_016869799.1
XM_017014311.1 Intron XP_016869800.1
XM_017014312.1 Intron XP_016869801.1

View Full Product Details