Product Details

SNP ID
rs6701146
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:65799816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGACTGTGCGAAATACAAACAGG[C/G]AACTCAGCCCACAGTGCTTGCTCTC
Phenotype
MIM: 600127
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDE4B PubMed Links

Gene Details

Gene
PDE4B
Gene Name
phosphodiesterase 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037339.2 Intron NP_001032416.1
NM_001037340.2 Intron NP_001032417.1
NM_001037341.1 Intron NP_001032418.1
NM_001297440.1 Intron NP_001284369.1
NM_001297441.1 Intron NP_001284370.1
NM_001297442.1 Intron NP_001284371.1
NM_002600.3 Intron NP_002591.2
XM_005270924.3 Intron XP_005270981.1
XM_005270925.2 Intron XP_005270982.1
XM_006710680.3 Intron XP_006710743.1
XM_011541566.1 Intron XP_011539868.1
XM_017001445.1 Intron XP_016856934.1
XM_017001446.1 Intron XP_016856935.1

View Full Product Details