Product Details

SNP ID
rs12571156
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:50802159 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTCCTTAGTTTCACTCATTATAG[C/T]GCAACTTACTTTGTTTTTCTAATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A1CF PubMed Links

Gene Details

Gene
A1CF
Gene Name
APOBEC1 complementation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198818.1 6612 UTR 3 NP_001185747.1
NM_001198819.1 6612 UTR 3 NP_001185748.1
NM_001198820.1 6612 UTR 3 NP_001185749.1
NM_014576.3 6612 UTR 3 NP_055391.2
NM_138932.2 6612 UTR 3 NP_620310.1
NM_138933.2 6612 UTR 3 NP_620311.1
XM_005269718.2 6612 UTR 3 XP_005269775.1
XM_005269720.3 6612 UTR 3 XP_005269777.1
XM_011539729.2 6612 UTR 3 XP_011538031.1
XM_011539730.2 6612 UTR 3 XP_011538032.1
XM_017016160.1 6612 UTR 3 XP_016871649.1

View Full Product Details