Product Details

SNP ID
rs12603404
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:44146546 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTAGAGTAAAGGAATCTGACAGCT[A/G]AAGTTTCTGTTCCTTGAAAATGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf53 PubMed Links

Gene Details

Gene
C17orf53
Gene Name
chromosome 17 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171251.2 Intron NP_001164722.1
NM_001321310.1 Intron NP_001308239.1
NM_001321311.1 Intron NP_001308240.1
NM_024032.4 Intron NP_076937.2
XM_011525187.1 Intron XP_011523489.1
XM_011525188.1 Intron XP_011523490.1
XM_011525189.1 Intron XP_011523491.1
XM_011525190.1 Intron XP_011523492.1
XM_011525191.2 Intron XP_011523493.1
XM_011525192.1 Intron XP_011523494.1
XM_011525193.1 Intron XP_011523495.1
XM_011525194.1 Intron XP_011523496.1
XM_011525195.1 Intron XP_011523497.1
XM_011525197.2 Intron XP_011523499.1
XM_017025031.1 Intron XP_016880520.1
XM_017025032.1 Intron XP_016880521.1

View Full Product Details