Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001005336.2 | Intron | NP_001005336.1 | |||
NM_001288737.1 | Intron | NP_001275666.1 | |||
NM_001288738.1 | Intron | NP_001275667.1 | |||
NM_001288739.1 | Intron | NP_001275668.1 | |||
NM_004408.3 | Intron | NP_004399.2 | |||
XM_005251763.2 | Intron | XP_005251820.1 | |||
XM_005251768.2 | Intron | XP_005251825.1 | |||
XM_005251769.2 | Intron | XP_005251826.1 | |||
XM_006716992.2 | Intron | XP_006717055.1 | |||
XM_006716993.1 | Intron | XP_006717056.1 | |||
XM_011518335.2 | Intron | XP_011516637.1 | |||
XM_011518336.2 | Intron | XP_011516638.1 | |||
XM_017014369.1 | Intron | XP_016869858.1 | |||
XM_017014370.1 | Intron | XP_016869859.1 | |||
XM_017014371.1 | Intron | XP_016869860.1 | |||
XM_017014372.1 | Intron | XP_016869861.1 | |||
XM_017014373.1 | Intron | XP_016869862.1 |