Product Details
- SNP ID
-
hCV11896232
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:55396542 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCCCGGGGTGGAGGGCGGTCGG[C/G]TGCGCTGTTTTCCGGTGAGGCCTGG
- Phenotype
-
MIM: 614726
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM165
PubMed Links
Gene Details
- Gene
- TMEM165
- Gene Name
- transmembrane protein 165
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018475.4 |
|
Intron |
|
|
NP_060945.2 |
View Full Product Details