Product Details

SNP ID
rs14580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82426604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATCCTTGATCATAATAGTTAT[A/T]AAATCCTTGGTTCCAGTTTTGGCCC
Phenotype
MIM: 607137
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ENOPH1 PubMed Links

Gene Details

Gene
ENOPH1
Gene Name
enolase-phosphatase 1
There are no transcripts associated with this gene.

Gene
HNRNPDL
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207000.1 1586 Intron NP_001193929.1
NM_031372.3 1586 Missense Mutation AAT,TAT N351Y NP_112740.1

View Full Product Details