Product Details

SNP ID
rs1820577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82357625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAATTCTTCAAATTAGAACACCCA[C/T]TCCATCTAAGGTCTTCACAACTATT
Phenotype
MIM: 601324
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPD PubMed Links

Gene Details

Gene
HNRNPD
Gene Name
heterogeneous nuclear ribonucleoprotein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003810.1 Intron NP_001003810.1
NM_002138.3 Intron NP_002129.2
NM_031369.2 Intron NP_112737.1
NM_031370.2 Intron NP_112738.1

View Full Product Details