Product Details

SNP ID
rs3821731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:21420517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGTTATATCTTCAAGCTAGA[C/T]AGGAGGGGAGAATGCTGACCAGGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF385D PubMed Links

Gene Details

Gene
ZNF385D
Gene Name
zinc finger protein 385D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024697.2 2497 Intron NP_078973.1
XM_011534122.1 2497 Intron XP_011532424.1
XM_011534123.2 2497 Intron XP_011532425.1
XM_011534124.2 2497 Intron XP_011532426.1
XM_017007191.1 2497 UTR 3 XP_016862680.1
XM_017007192.1 2497 UTR 3 XP_016862681.1
XM_017007193.1 2497 UTR 3 XP_016862682.1
XM_017007194.1 2497 UTR 3 XP_016862683.1
XM_017007195.1 2497 UTR 3 XP_016862684.1
XM_017007196.1 2497 UTR 3 XP_016862685.1
XM_017007197.1 2497 UTR 3 XP_016862686.1
XM_017007198.1 2497 UTR 3 XP_016862687.1
XM_017007199.1 2497 UTR 3 XP_016862688.1
XM_017007200.1 2497 UTR 3 XP_016862689.1
XM_017007201.1 2497 UTR 3 XP_016862690.1
XM_017007202.1 2497 UTR 3 XP_016862691.1
XM_017007203.1 2497 Intron XP_016862692.1

View Full Product Details