Product Details

SNP ID
rs1792833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118367281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCGAAGGAATTTCATCAATAA[A/G]AAAAAAATAACAAGAAAGAATAAAA
Phenotype
MIM: 603753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE4A PubMed Links

Gene Details

Gene
UBE4A
Gene Name
ubiquitination factor E4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204077.1 Intron NP_001191006.1
NM_004788.3 Intron NP_004779.2

View Full Product Details