Product Details

SNP ID
rs13770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119419060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCCTCCAGTGGCTGGTACCCCA[A/C]CATCCCACTACCCCTCACATGCTCT
Phenotype
MIM: 188230
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
THY1 PubMed Links

Gene Details

Gene
THY1
Gene Name
Thy-1 cell surface antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311160.1 1874 UTR 3 NP_001298089.1
NM_001311162.1 1874 UTR 3 NP_001298091.1
NM_006288.4 1874 UTR 3 NP_006279.2
Gene
USP2-AS1
Gene Name
USP2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details