Product Details

SNP ID
rs1860849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190654134 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTGAAACTTTGCTAGGAATAAAT[C/G]GGGATAAAAGAGTAATGAAACCTAA
Phenotype
MIM: 600800
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NAB1 PubMed Links

Gene Details

Gene
NAB1
Gene Name
NGFI-A binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321312.1 Intron NP_001308241.1
NM_001321313.1 Intron NP_001308242.1
NM_001321314.1 Intron NP_001308243.1
NM_001321315.1 Intron NP_001308244.1
NM_005966.3 Intron NP_005957.2
XM_005246582.1 Intron XP_005246639.1
XM_005246583.1 Intron XP_005246640.1
XM_005246586.1 Intron XP_005246643.1
XM_005246587.1 Intron XP_005246644.1
XM_011511219.2 Intron XP_011509521.1
XM_017004170.1 Intron XP_016859659.1
XM_017004171.1 Intron XP_016859660.1
XM_017004172.1 Intron XP_016859661.1
XM_017004173.1 Intron XP_016859662.1
XM_017004174.1 Intron XP_016859663.1
XM_017004175.1 Intron XP_016859664.1
XM_017004176.1 Intron XP_016859665.1

View Full Product Details