Product Details

SNP ID
rs2073307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:68875533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTTAAGCAGCTTTGCAGAAATG[C/T]GGGCCTTGCCCCATGCAAGCAGCTC
Phenotype
MIM: 102575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTN1 PubMed Links

Gene Details

Gene
ACTN1
Gene Name
actinin alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102.3 Intron NP_001093.1
NM_001130004.1 Intron NP_001123476.1
NM_001130005.1 Intron NP_001123477.1
XM_011537265.2 Intron XP_011535567.1
XM_011537266.2 Intron XP_011535568.1
XM_011537267.2 Intron XP_011535569.1
XM_011537268.2 Intron XP_011535570.1
XM_017021720.1 Intron XP_016877209.1
XM_017021721.1 Intron XP_016877210.1
XM_017021722.1 Intron XP_016877211.1
XM_017021723.1 Intron XP_016877212.1
XM_017021725.1 Intron XP_016877214.1
XM_017021726.1 Intron XP_016877215.1
XM_017021727.1 Intron XP_016877216.1
XM_017021728.1 Intron XP_016877217.1

View Full Product Details