Product Details
- SNP ID
-
rs1888823
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35300633 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCTTGTAGAAAGTACAATGGCTTC[A/G]TCCAGTTGGGTTGAGAACAGAGAAA
- Phenotype
-
MIM: 610094
MIM: 194549
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DEF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74336218] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEF6
- Gene Name
- DEF6, guanine nucleotide exchange factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022047.3 |
|
Intron |
|
|
NP_071330.3 |
- Gene
- ZNF76
- Gene Name
- zinc finger protein 76
There are no transcripts associated with this gene.
View Full Product Details