Product Details

SNP ID
rs1050827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154546123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTTCCCGCAGGATCCCGCACAC[G/C]TGGGTCCGGCTCAGGGCCACCTGCT
Phenotype
MIM: 305900 MIM: 300248
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 160 Missense Mutation CAC,CAG H41Q NP_000393.4
NM_001042351.2 160 Missense Mutation CAC,CAG H11Q NP_001035810.1
Gene
IKBKG
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099856.4 160 Intron NP_001093326.2
NM_001099857.2 160 Intron NP_001093327.1
NM_001145255.2 160 Intron NP_001138727.1
NM_001321396.1 160 Intron NP_001308325.1
NM_001321397.1 160 Intron NP_001308326.1
NM_003639.4 160 Intron NP_003630.1

View Full Product Details