Product Details
- SNP ID
-
rs1865434
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:109756261 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGCTGAGCGTCTTCTTTTAATGATA[C/T]TCTCTGACATGTGACATCCTGGTGA
- Phenotype
-
MIM: 600797
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IRS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74123739] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IRS2
- Gene Name
- insulin receptor substrate 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003749.2 |
4574 |
UTR 3 |
|
|
NP_003740.2 |
View Full Product Details