Product Details
- SNP ID
-
rs1885988
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:27436125 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAATTTTAAATCTCCACTTCGTCA[C/T]GGCACGAAGAGGCTACGTTTGGCTC
- Phenotype
-
MIM: 600860
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GTF3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146301498] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF3A
- Gene Name
- general transcription factor IIIA
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002097.2 |
|
Intron |
|
|
NP_002088.2 |
- Gene
- MTIF3
- Gene Name
- mitochondrial translational initiation factor 3
View Full Product Details