Product Details
- SNP ID
-
rs917593
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40660274 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTGTTTCTGAAATCTGATTTGCA[A/G]TATCTTCATCTTTTTTTTTTTTTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT222
PubMed Links
Gene Details
- Gene
- KRT222
- Gene Name
- keratin 222
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152349.2 |
|
Intron |
|
|
NP_689562.1 |
View Full Product Details